5-Chloro-2,2-difluoro-1,3-benzodioxole ≥97%

Supplier: AOBChem USA

Warning

121819-5G 121819-25G
77359-872EA 133.9 USD
77359-872 77371-860
5-Chloro-2,2-difluoro-1,3-benzodioxole ≥97%
5-Chloro-2,2-difluoro-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972706
CAS Number: 72769-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR