4-Bromo-2-methyl-5-nitroaniline ≥97%

Supplier: AOBChem USA

Warning

120740-5G 120740-25G
77359-854EA 148.32 USD
77359-854 77371-842
4-Bromo-2-methyl-5-nitroaniline ≥97%
4-Bromo-2-methyl-5-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03942039
CAS Number: 71785-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR