Methyl 3-chloro-4-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

120633-5G 120633-25G
77359-844EA 174.07 USD
77359-844 77371-834
Methyl 3-chloro-4-(trifluoromethyl)benzoate ≥95%
Methyl 3-chloro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698014
CAS Number: 1214345-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR