1-(4-Bromophenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

120607-5G 120607-25G
77359-838EA 151.41 USD
77359-838 77371-826
1-(4-Bromophenyl)propan-1-ol ≥97%
1-(4-Bromophenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD00021911
CAS Number: 4489-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR