Methyl 2-bromo-3-chlorobenzoate ≥95%

Supplier: AOBChem USA

Warning

120594-5G 120594-25G
77359-836EA 129.78 USD
77359-836 77371-824
Methyl 2-bromo-3-chlorobenzoate ≥95%
Methyl 2-bromo-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398719
CAS Number: 1107627-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR