4-Cyclopentylphenol ≥95%

Supplier: AOBChem USA

Warning

119195-5G 119195-25G
77359-792EA 131.84 USD
77359-792 77371-784
4-Cyclopentylphenol ≥95%
4-Cyclopentylphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00009953
CAS Number: 1518-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR