(6-Bromo-3-ethoxy-2-fluorophenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118167-5G 118167-25G
77359-764EA 161.71 USD
77359-764 77371-750
(6-Bromo-3-ethoxy-2-fluorophenyl)(phenyl)methanol ≥97%
(6-Bromo-3-ethoxy-2-fluorophenyl)(phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFO₂
MW: 325.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783923
CAS Number: 2901099-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR