3-Bromo-5-(tert-butyl)toluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-(tert-butyl)-5-methylbenzene

116447-5G 116447-25G
77359-686EA 152.44 USD
77359-686 77371-684
3-Bromo-5-(tert-butyl)toluene ≥97%
3-Bromo-5-(tert-butyl)toluene

Precaución: for Research Use Only

Formula: C₁₁H₁₅Br
MW: 227.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801037
CAS Number: 193905-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR