3-Chloro-2,6-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-2,6-difluorophenyl)boronic acid

11452-5G 11452-25G
77359-658EA 88.58 USD
77359-658 77371-648
3-Chloro-2,6-difluorophenylboronic acid ≥97%
3-Chloro-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04038753
CAS Number: 1031226-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR