2,6-Diethylphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Diethyl-2-hydroxybenzene

114415-5G 114415-25G
77359-652EA 107.12 USD
77359-652 77371-642
2,6-Diethylphenol ≥97%
2,6-Diethylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD01707589
CAS Number: 1006-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR