5-Bromo-2-(trifluoromethoxy)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-cyano-1-(trifluoromethoxy)benzene

113687-5G 113687-25G
77359-636EA 60.77 USD
77359-636 77371-628
5-Bromo-2-(trifluoromethoxy)benzonitrile ≥95%
5-Bromo-2-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD13185875
CAS Number: 1210906-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR