3-Chloro-4-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(trifluoromethyl)benzeneboronic acid

11282-5G 11282-25G
77359-614EA 75.19 USD
77359-614 77371-590
3-Chloro-4-(trifluoromethyl)phenylboronic acid ≥95%
3-Chloro-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788417
CAS Number: 847756-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR