2-Fluoro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

112396-5G 112396-25G
77359-604EA 154.5 USD
77359-604 77371-582
2-Fluoro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide ≥97%
2-Fluoro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD34781535
CAS Number: 2901096-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR