4-Bromo-2-((2-fluorobenzyl)oxy)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

110439-5G 110439-25G
77359-580EA 100.94 USD
77359-580 77371-552
4-Bromo-2-((2-fluorobenzyl)oxy)-1-methylbenzene ≥97%
4-Bromo-2-((2-fluorobenzyl)oxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780611
CAS Number: 1986772-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR