2-Bromo-4-(tert-butyl)-1-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

109819-5G 109819-25G
77359-568EA 130.81 USD
77359-568 77371-536
2-Bromo-4-(tert-butyl)-1-fluorobenzene ≥95%
2-Bromo-4-(tert-butyl)-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrF
MW: 231.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845933
CAS Number: 34252-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR