3-Bromo-2,6-bis(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

107298-5G 107298-25G
77359-506EA 162.74 USD
77359-506 77371-468
3-Bromo-2,6-bis(methylthio)pyridine ≥95%
3-Bromo-2,6-bis(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNS₂
Storage Temperature: Ambient
CAS Number: 1478090-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR