2-Bromo-1-ethoxy-3,5-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

105169-5G 105169-25G
77359-474EA 182.31 USD
77359-474 77371-448
2-Bromo-1-ethoxy-3,5-difluorobenzene ≥95%
2-Bromo-1-ethoxy-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32705920
CAS Number: 2386931-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR