N,N-Diethylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

102940-5G 102940-25G
77359-418EA 60.77 USD
77359-418 77371-402
N,N-Diethylisonicotinamide ≥97%
N,N-Diethylisonicotinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00059785
CAS Number: 530-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR