1-(5-Ethoxy-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

102649-5G 102649-25G
77359-412EA 154.5 USD
77359-412 77371-398
1-(5-Ethoxy-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%
1-(5-Ethoxy-2-fluorophenyl)-4,4-dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₆H₂₃FO₂
MW: 266.351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777234
CAS Number: 2901103-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR