2-Bromo-6-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

101302-5G 101302-25G
77359-382EA 176.13 USD
77359-382 77371-370
2-Bromo-6-fluoro-N-methoxy-N-methylbenzamide ≥97%
2-Bromo-6-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD21333087
CAS Number: 1341037-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR