6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

101217-5G 101217-25G
77359-380EA 131.84 USD
77359-380 77371-368
6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid ≥97%
6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21365092
CAS Number: 1026962-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR