1-Benzyl-1H-pyrazole-4-boronic acid

Supplier: AOBChem USA

Warning

11071-1G 11071-25G 11071-5G
76882-104EA 68.96 USD
76882-104 76882-106 76882-108
1-Benzyl-1H-pyrazole-4-boronic acid
1-Benzyl-1H-pyrazole-4-boronic acid

for Research Use Only

Formula: C₁₀H₁₁BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04038760
CAS Number: 852362-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR