Brady® HH83 and HH85 RFID Reader Accessories
Supplier: Brady Worldwide
176284 311014 311015 311031 311034
77322-098EA 92.03 USD
77322-098 77322-106 77322-108 77322-110 77322-112 77322-114
Brady® HH83 and HH85 RFID Reader Accessories
Barcode and RFID Systems
Keep workflow interruptions to a minimum with long-lasting battery and battery chargers for HH83 and HH85. Use holster to carry your HH83 reader comfortable and conveniently.
  • Batteries, battery chargers, and holster for use with HH83 and/or HH85 RFID readers
HH83 and HH85 4-bay charger - Features 4 bays to charge 4 HH83/HH85 readers, plus 4 additional battery charging slots. Provides a dedicated charging location for handheld device fleet
HH83 barcode scanner battery - 3350 mAh battery includes a cover. Use with the HH83 Handheld Barcode Scanner and mobile computer (not recommended for the HH83 UHF RFID Reader and mobile computer).
HH83 barcode scanner and RFID reader battery - 6700mAh battery includes a cover. Use with the HH83 Handheld Barcode Scanner and mobile computer or the HH83 UHF RFID Reader and mobile computer.
HH83 and HH85 single bay charger - Features a single bay to charge 1 HH83/HH85 reader, plus has 1 spare battery charging slot. Provides a dedicated charging location for handheld device fleet.
HH83 and HH85 battery with pistol grip - 10050 mAh battery for any HH Series device. Direct backup or replacement for HH85 series battery and pistol grip. Use with an HH83 device to add a pistol grip with trigger for additional functionality and to expand battery life.
HH83 holster - Flexible and durable nylon web construction. Accommodates belts up to 3" wide. Use with HH83 Series devices.
Información de suministro: Lithium-ion batteries must ship ground.