Methyl 2-methyl-3-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

62354-5G 62354-25G
77355-860EA 60.77 USD
77355-860 77379-904
Methyl 2-methyl-3-(trifluoromethyl)benzoate ≥95%
Methyl 2-methyl-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14698066
CAS Number: 346603-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR