3-Bromo-2-chloro-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49381-5G 49381-25G
77351-462EA 60.77 USD
77351-462 77375-750
3-Bromo-2-chloro-5-fluorobenzoic acid ≥97%
3-Bromo-2-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28166248
CAS Number: 1805210-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR