3-Bromo-5-chloro-tert-butylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-(tert-butyl)-5-chlorobenzene

47148-5G 47148-25G
77350-864EA 60.77 USD
77350-864 77375-176
3-Bromo-5-chloro-tert-butylbenzene ≥97%
3-Bromo-5-chloro-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrCl
MW: 247.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089328
CAS Number: 1263377-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR