3-Chlorovanillin ≥95%

Supplier: AOBChem USA

Warning

46123-5G 46123-25G
77350-642EA 60.77 USD
77350-642 77374-898
3-Chlorovanillin ≥95%
3-Chlorovanillin

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016982
CAS Number: 19463-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR