3-Bromo-9-isopropyl-9H-carbazole ≥97%

Supplier: AOBChem USA

Warning

38044-5G 38044-25G
77350-274EA 60.77 USD
77350-274 77374-458
3-Bromo-9-isopropyl-9H-carbazole ≥97%
3-Bromo-9-isopropyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrN
MW: 288.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529942
CAS Number: 55141-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR