3-Chloropyridine-4-carboxylic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 3-chloroisonicotinate

29108-5G 29108-25G
77348-796EA 60.77 USD
77348-796 77373-060
3-Chloropyridine-4-carboxylic acid ethyl ester ≥97%
3-Chloropyridine-4-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411706
CAS Number: 211678-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR