4-Trifluoromethoxythioanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl(4-(trifluoromethoxy)phenyl)sulphane, 4-(Trifluoromethoxy)phenyl methyl sulphphide, 4-(Trifluoromethoxy)phenyl methyl sulphide

17710-5G 17710-25G
77348-108EA 60.77 USD
77348-108 77372-332
4-Trifluoromethoxythioanisole ≥95%
4-Trifluoromethoxythioanisole

Precaución: for Research Use Only

Formula: C₈H₇F₃OS
MW: 208.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320776
CAS Number: 2546-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR