2-Chloro-4-fluoro-5-nitrobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15233-5G 15233-25G
77347-992EA 60.77 USD
77347-992 77372-182
2-Chloro-4-fluoro-5-nitrobenzaldehyde ≥95%
2-Chloro-4-fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₃
Storage Temperature: Ambient
MDL Number: MFCD17168722
CAS Number: 99329-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR