2-Butoxy-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14417-5G 14417-25G
77347-946EA 60.77 USD
77347-946 77372-106
2-Butoxy-6-fluorophenylboronic acid ≥97%
2-Butoxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
Storage Temperature: Ambient
MDL Number: MFCD06798079
CAS Number: 870777-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR