5-Methylbenzo[b]thiophene-2-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
99994-1G
99994-5G
99994-25G
77341-472EA
88.58
USD
77341-472
77371-358
77383-352
5-Methylbenzo[b]thiophene-2-carboxylic acid ≥97%
5-Methylbenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈O₂S MW: 192.234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04969088 CAS Number: 1505-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...