Methyl 5-methylbenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99993-500MG 99993-1G 99993-5G
77335-596EA 359.47 USD
77335-596 77359-364 77383-350
Methyl 5-methylbenzo[b]thiophene-2-carboxylate ≥97%
Methyl 5-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD11044569
CAS Number: 82787-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR