4-(3-(tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

99382-500MG 99382-1G 99382-5G 99382-25G
77335-276EA 243.08 USD
77335-276 77359-358 77371-350 77391-762
4-(3-(tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(3-(tert-butyl)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₅H₂₂O₂
MW: 234.334 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21090819
CAS Number: 1353499-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR