4-(2-Isopropylphenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

99350-500MG 99350-1G 99350-5G 99350-25G
77335-272EA 264.71 USD
77335-272 77359-354 77371-346 77391-758
4-(2-Isopropylphenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(2-Isopropylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MW: 220.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31694052
CAS Number: 1869788-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR