1-(3-Bromo-2-fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

99331-500MG 99331-1G 99331-5G 99331-25G
77335-266EA 182.31 USD
77335-266 77347-068 77371-342 77391-752
1-(3-Bromo-2-fluorophenyl)cyclopentanol ≥95%
1-(3-Bromo-2-fluorophenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO
MW: 259.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34775966
CAS Number: 1504737-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR