1-(6-Bromopyridin-2-yl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

99316-500MG 99316-1G 99316-5G
77339-738EA 621.09 USD
77339-738 77359-348 77383-342
1-(6-Bromopyridin-2-yl)-2-fluoroethanone ≥95%
1-(6-Bromopyridin-2-yl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
Storage Temperature: Ambient
CAS Number: 1093880-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR