1-Bromo-4-chloro-3-fluoro-2-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

99178-500MG 99178-1G 99178-5G 99178-25G
77335-264EA 184.37 USD
77335-264 77347-350 77371-340 77391-750
1-Bromo-4-chloro-3-fluoro-2-isopropoxybenzene ≥95%
1-Bromo-4-chloro-3-fluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34775893
CAS Number: 2918917-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR