Methyl 4,6-dibromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99174-500MG 99174-1G 99174-5G 99174-25G
77335-262EA 158.62 USD
77335-262 77345-732 77371-338 77391-748
Methyl 4,6-dibromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 4,6-dibromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂O₂S
Storage Temperature: Ambient
CAS Number: 2384437-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR