Methyl 4-chloro-7-methylbenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99162-1G 99162-5G 99162-25G
77340-856EA 60.77 USD
77340-856 77371-336 77383-338
Methyl 4-chloro-7-methylbenzo[b]thiophene-2-carboxylate ≥97%
Methyl 4-chloro-7-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉ClO₂S
MW: 240.706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044575
CAS Number: 1029720-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR