Methyl 4,7-dibromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99041-1G 99041-5G 99041-25G
77341-686EA 99.91 USD
77341-686 77371-332 77383-332
Methyl 4,7-dibromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 4,7-dibromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂O₂S
Storage Temperature: Ambient
CAS Number: 1126634-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR