4-Bromo-6-chlorobenzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

99036-500MG 99036-1G 99036-5G 99036-25G
77335-260EA 212.18 USD
77335-260 77359-344 77371-330 77391-746
4-Bromo-6-chlorobenzo[b]thiophene-2-carboxylic acid ≥97%
4-Bromo-6-chlorobenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₄BrClO₂S
Storage Temperature: Ambient
CAS Number: 1936547-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR