Methyl 4-bromo-6-chlorobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

99035-500MG 99035-1G 99035-5G 99035-25G
77335-258EA 182.31 USD
77335-258 77347-066 77371-328 77391-744
Methyl 4-bromo-6-chlorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 4-bromo-6-chlorobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₆BrClO₂S
Storage Temperature: Ambient
MDL Number: MFCD29423303
CAS Number: 2169644-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR