Methyl 4,5-difluorobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99029-500MG 99029-1G 99029-5G 99029-25G
77335-256EA 276.04 USD
77335-256 77359-342 77371-326 77391-742
Methyl 4,5-difluorobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 4,5-difluorobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD29378737
CAS Number: 2113523-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR