2-Chloro-6-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

98866-500MG 98866-1G 98866-5G 98866-25G
77335-254EA 109.18 USD
77335-254 77343-250 77371-322 77383-326
2-Chloro-6-(trifluoromethoxy)benzoic acid ≥97%
2-Chloro-6-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₃
MW: 240.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393799
CAS Number: 1261617-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR