6-(4-Chlorophenoxy)nicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

98841-500MG 98841-1G 98841-5G 98841-25G
77335-250EA 256.47 USD
77335-250 77359-338 77371-318 77391-738
6-(4-Chlorophenoxy)nicotinaldehyde ≥95%
6-(4-Chlorophenoxy)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO₂
MW: 233.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12403521
CAS Number: 1160430-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR