6-(4-Chlorophenoxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

98839-500MG 98839-1G 98839-5G
77335-248EA 317.24 USD
77335-248 77359-336 77383-324
6-(4-Chlorophenoxy)nicotinic acid ≥97%
6-(4-Chlorophenoxy)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD06208648
CAS Number: 51362-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR