2-Iodo-6-methoxy-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

98716-500MG 98716-1G 98716-5G 98716-25G
77335-244EA 89.61 USD
77335-244 77342-354 77371-312 77383-316
2-Iodo-6-methoxy-4-methylphenol ≥97%
2-Iodo-6-methoxy-4-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉IO₂
MW: 264.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527540
CAS Number: 111726-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR