Methyl-4-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-(methylthio)benzoate

98584-1G 98584-5G 98584-25G
77341-972EA 109.18 USD
77341-972 77371-310 77383-314
Methyl-4-(methylthio)benzoate ≥95%
Methyl 4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
MW: 182.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01313010
CAS Number: 3795-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR